missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane, 99.5%, Extra Dry over Molecular Sieve, stabilized, AcroSeal™, Thermo Scientific Chemicals

Catalog Number 15214228
45.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


42.30 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.


Click to view available options
Quantity:
500 mL

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 12.0°C
CAS 123-91-1
Density 1.0340g/mL
Boiling Point 101.0°C
Quantity 500 mL
Flash Point 12°C
Refractive Index 1.4205 to 1.4220
Specific Gravity 1.034
Molecular Formula C4H8O2
Viscosity 1.32 mPa.s (20°C)
Water 0.005% max.
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone, benzene, toluene,, ether and other common organic solvents
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
SMILES C1COCCO1
IUPAC Name 1,4-dioxane
Molecular Weight (g/mol) 88.106
PubChem CID 31275
ChEBI CHEBI:47032
Formula Weight 88.11
Percent Purity 99.5%
Grade Extra Dry over Molecular Sieve
Chemical Name or Material 1,4-Dioxane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,4-Dioxane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'