missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane-2,5-dione, TRC

Product Code. 30383258
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383258 1 g
30386122 100 mg
30373821 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383258 Supplier Toronto Research Chemicals Supplier No. TRCD4858831G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 502-97-6
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
Synonym Glycolide S, Glycollide, Glycolide
SMILES O=C1COC(=O)CO1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C4H4O4/c5-3-1-7-4(6)2-8-3/h1-2H2
Formula Weight 116.011
Chemical Name or Material 1,4-Dioxane-2,5-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.