missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane-2,3-diol, 98%

Product Code. 11916211 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11916211 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11916211 Supplier Thermo Scientific Acros Supplier No. 438370050
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4845-50-5 | C4H8O4 | 120.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 92°C to 94°C
CAS 4845-50-5
CAS Min % 97.5
Color White
CAS Max % 100.0
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Powder or Crystalline Powder
Assay Percent Range 98%
Molecular Formula C4H8O4
MDL Number MFCD00006572
Synonym 2,3-dihydroxy-1,4-dioxane, p-dioxane-2,3-diol, 2,3-dihydroxy-1,4-dioxan, dioxanediol, 1,3-diol, 1,4dioxane-2,3-diol, acmc-1al8g, dsstox_cid_18824, dsstox_rid_79404, dsstox_gsid_38824
InChI Key YLVACWCCJCZITJ-UHFFFAOYNA-N
SMILES OC1OCCOC1O
IUPAC Name 1,4-dioxane-2,3-diol
Molecular Weight (g/mol) 120.10
PubChem CID 96170
Formula Weight 120.11
Percent Purity 98%
Chemical Name or Material 1, 4-Dioxane-2, 3-diol
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Dioxane-2,3-diol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.