missing translation for 'onlineSavingsMsg'
Learn More

Riedel-de Haën 1,4-Dioxane

Product Code. 15925660
Change view
Click to view available options
Quantity:
7 L
Unit Size:
7 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15925660 7 L 7 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15925660 Supplier Riedel-de Haën Supplier No. 331477L
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

1,4-Dioxane, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5% (GC).

  • ACS general use solvents.
  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis.
  • Suitable for less demanding applications such as routine isocratic HPLC.
  • Variations and decrease issues traceable to solvent contamination.
  • All solvents are tested to meet ACS general use specifications.

Specifications

pH 6 to 8 at 500 g/l (20°C)
Melting Point 12°C
CAS 123-91-1
Color Colorless
CAS Max % ≤100.00%
Density 1.031 to 1.034 g/cm3 (20°C)
Quantity 7 L
Flash Point 22.5 %(V)
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line Puriss. p.a.
Molecular Formula C4H8O2
Linear Formula C4H8O2
Vapor Pressure 36 hPa (20°C)
MDL Number MFCD00006571
Viscosity 1.27 mPa.s (20°C)
UN Number UN1165
Beilstein 102551
Synonym Diethylene oxide, Dioxane
Boiling Point 100°C to 102°C (1.013 hPa)
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
SMILES C1COCCO1
IUPAC Name 1,4-dioxane
Molecular Weight (g/mol) 88.11
Percent Purity ≥99.50%
Grade ACS
Chemical Name or Material 1,4-Dioxane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.