missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dioxan Anhydrous, Honeywell™ Riedel de-Haen
Description
- These solvents are specially produced to a water specification of less than 50 ppm (<0.0050%).
- Many solvents go well beyond that and are routinely produced to lower than 5 ppm (<0.0005%).
- All packaging steps and materials are especially designed to retain solvent dryness and ensure product integrity. Before filling, amber glass bottles are oven-dried to remove all traces of water, then filled under dry nitrogen and sealed with a special cap.
- The water content itself is strictly controlled by coulometric Karl Fischer analysis (Hydranal™) and actual results are printed on the label.
- This line is packaged in specially designed septum-sealed bottles to maintain low water content and ensure product integrity
Specifications
Specifications
| Density | 1.03 g/cm3 (20°C) |
| Quantity | 100 mL |
| Flash Point | 11°C |
| Packaging | Septum Sealed Bottle |
| UN Number | 1165 |
| Formula Weight | 88.11 |
| Percent Purity | >99.9% |
| Chemical Name or Material | 1,4-Dioxan |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?