missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxa-8-azaspiro[4.5]decane, 98%

Product Code. 10419650 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10419650 50 g Glass bottle 50g
10489480 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10419650 Supplier Thermo Scientific Acros Supplier No. 176370500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 177-11-7 | C7H14NO2 | 144.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177-11-7
Molecular Formula C7H14NO2
Molecular Weight (g/mol) 144.19
MDL Number MFCD00005976
InChI Key KPKNTUUIEVXMOH-UHFFFAOYSA-O
Synonym 1,4-dioxa-8-azaspiro 4.5 decane, 4-piperidone ethylene ketal, 4-piperidinone ethyl ketal, 4-piperidone-ethylene ketal, 4-piperidone ethylene acetal, 1,4-dioxa-8-azaspiro 4,5 decane, 4-piperidoneethyleneketal, piperidone-4-ethyleneketal, 1,4-dioxa-8-aza-spiro 4.5 decane
PubChem CID 67435
IUPAC Name 1,4-dioxa-8-azaspiro[4.5]decane
SMILES C1COC2(CC[NH2+]CC2)O1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1100g/mL
Boiling Point 108.0°C to 110.0°C (26.0 mmHg)
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4809 to 1.4829
Quantity 50 g
Specific Gravity 1.11
Formula Weight 143.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,4-Dioxa-8-azaspiro[4.5]decane, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Dioxa-8-azaspiro¢4.5!decane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.