missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dinitrobenzene, 98+%, Thermo Scientific Chemicals

Catalog Number 11424270
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
100-25-4
C6H4N2O4
168.108
MFCD00007314
FYFDQJRXFWGIBS-UHFFFAOYSA-N
p-dinitrobenzene, benzene, 1,4-dinitro, benzene, p-dinitro, dithane a-4, dinitrobenzene, p, dinitrobenzene, para, ccris 3092, 1,4-dinitro-benzene, 1,4-dinitrobenzene, analytical standard, para-dinitrobenzene
7492
CHEBI:51398
1,4-dinitrobenzene
C1=CC(=CC=C1[N+](=O)[O-])[N+](=O)[O-]
This item is not returnable. View return policy
100-25-4
C6H4N2O4
168.108
MFCD00007314
FYFDQJRXFWGIBS-UHFFFAOYSA-N
p-dinitrobenzene, benzene, 1,4-dinitro, benzene, p-dinitro, dithane a-4, dinitrobenzene, p, dinitrobenzene, para, ccris 3092, 1,4-dinitro-benzene, 1,4-dinitrobenzene, analytical standard, para-dinitrobenzene
7492
CHEBI:51398
1,4-dinitrobenzene
C1=CC(=CC=C1[N+](=O)[O-])[N+](=O)[O-]

1,4-Dinitrobenzene is used in a study to evaluate the ionization mechanism and solvent effect by novel atmospheric pressure photoionization mass spectrometry in negative ion mode for analysis of some compounds. 1,4-Dinitrobenzene can be used in synthesis of dyes and dye intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dinitrobenzene is used in a study to evaluate the ionization mechanism and solvent effect by novel atmospheric pressure photoionization mass spectrometry in negative ion mode for analysis of some compounds. 1,4-Dinitrobenzene can be used in synthesis of dyes and dye intermediates.

Solubility
Soluble in water. (0.8 g/L) at 20°C.

Notes
Incompatible with bases, alkali metals, oxidizing agents and nitric acid.
TRUSTED_SUSTAINABILITY
Melting Point 172°C to 176°C
Density 1.625
Boiling Point 299°C
Flash Point 150°C (302°F)
Quantity 100 g
UN Number UN3443
Beilstein 1105828
Merck Index 14,3273
Solubility Information Soluble in water. (0.8g/L) at 20°C.
Formula Weight 168.11
Percent Purity ≥98%
Chemical Name or Material 1,4-Dinitrobenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'