missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dinitrobenzene, 98+%

Product Code. 11414270
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414270 25 g 25g
11404270 5 g 5g
11424270 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414270 Supplier Thermo Scientific Alfa Aesar Supplier No. A10293.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-25-4 | C6H4N2O4 | 168.108 g/mol

1,4-Dinitrobenzene is used in a study to evaluate the ionization mechanism and solvent effect by novel atmospheric pressure photoionization mass spectrometry in negative ion mode for analysis of some compounds. 1,4-Dinitrobenzene can be used in synthesis of dyes and dye intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dinitrobenzene is used in a study to evaluate the ionization mechanism and solvent effect by novel atmospheric pressure photoionization mass spectrometry in negative ion mode for analysis of some compounds. 1,4-Dinitrobenzene can be used in synthesis of dyes and dye intermediates.

Solubility
Soluble in water. (0.8 g/L) at 20°C.

Notes
Incompatible with bases, alkali metals, oxidizing agents and nitric acid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-25-4
Molecular Formula C6H4N2O4
Molecular Weight (g/mol) 168.108
MDL Number MFCD00007314
InChI Key FYFDQJRXFWGIBS-UHFFFAOYSA-N
Synonym p-dinitrobenzene, benzene, 1,4-dinitro, benzene, p-dinitro, dithane a-4, dinitrobenzene, p, dinitrobenzene, para, ccris 3092, 1,4-dinitro-benzene, 1,4-dinitrobenzene, analytical standard, para-dinitrobenzene
PubChem CID 7492
ChEBI CHEBI:51398
IUPAC Name 1,4-dinitrobenzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 172°C to 176°C
Density 1.625
Boiling Point 299°C
Flash Point 150°C (302°F)
Quantity 25 g
UN Number UN3443
Beilstein 1105828
Merck Index 14,3273
Solubility Information Soluble in water. (0.8g/L) at 20°C.
Formula Weight 168.11
Percent Purity ≥98%
Chemical Name or Material 1,4-Dinitrobenzene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,4-Dinitrobenzene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 1,4-Dinitrobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.