missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethylpiperazine, 98%

Product Code. 11445877
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445877 100 g 100g
11435877 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445877 Supplier Thermo Scientific Alfa Aesar Supplier No. B24419.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 106-58-1 | C6H14N2 | 114.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-58-1
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00006156
InChI Key RXYPXQSKLGGKOL-UHFFFAOYSA-N
Synonym n,n'-dimethylpiperazine, lupetazine, piperazine, 1,4-dimethyl, texacat dmp, unii-tf146u602l, ccris 6690, 1,4-dimethyl-piperazine, 1,4-dimethyl-2,3,5,6-tetrahydropyrazine, 1,4-dimethylpiperazin, pubchem8569
PubChem CID 7818
IUPAC Name 1,4-dimethylpiperazine
SMILES CN1CCN(CC1)C

Specifications

Melting Point -1°C
Density 0.844
pH 10
Boiling Point 131°C to 132°C
Flash Point 21°C (69°F)
Odor Amine-like
Refractive Index 1.447
Quantity 100 g
UN Number UN2733
Beilstein 103022
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material 1,4-Dimethylpiperazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.