missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethyl-2,5-bis(phenylmethoxy)-benzene, TRC

Product Code. 30379154
Change view
Click to view available options
Quantity:
2.5 g
5 g
Unit Size:
2.50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30379154 5 g 5g
30384545 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30379154 Supplier Toronto Research Chemicals Supplier No. TRCD4789735G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,4-Dimethyl-2,5-bis(phenylmethoxy)-benzene
CAS 873973-47-8
Molecular Formula C13 H13 N O4 S . C13 H11 N O6 S
Quantity 5 g
Synonym 2,5-Bis(benzyloxy)-p-xylene
InChI Formula InChI=1S/C13H11NO6S.C13H13NO4S/c14-8-5-10(12(15)11(6-8)13(16)17)7-1-3-9(4-2-7)21(18,19)20;1-8-6-10(14)7-12(13(8)15)9-2-4-11(5-3-9)19(16,17)18/h1-6,15H,14H2,(H,16,17)(H,18,19,20);2-7,15H,14H2,1H3,(H,16,17,18)
SMILES Cc1cc(N)cc(c1O)c2ccc(cc2)S(=O)(=O)O.Nc3cc(C(=O)O)c(O)c(c3)c4ccc(cc4)S(=O)(=O)O
IUPAC Name 4-(5-amino-2-hydroxy-3-methylphenyl)benzenesulfonic acid;5-amino-2-hydroxy-3-(4-sulfophenyl)benzoic acid
Molecular Weight (g/mol) 588.606
Formula Weight 588.087
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.