missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethyl-2-(4-phenylbutoxy)benzene, TRC

Product Code. 30382337
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382337 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382337 Supplier Toronto Research Chemicals Supplier No. TRCD47671050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,4-Dimethyl-2-(4-phenylbutoxy)benzene
CAS 500904-64-3
Molecular Formula C18 H22 O
Quantity 50 mg
Synonym Benzene, 1,4-dimethyl-2-(4-phenylbutoxy)-, 1,4-Dimethyl-2-(4-phenylbutoxy)benzene
InChI Formula InChI=1S/C18H22O/c1-15-11-12-16(2)18(14-15)19-13-7-6-10-17-8-4-3-5-9-17/h3-5,8-9,11-12,14H,6-7,10,13H2,1-2H3
SMILES Cc1ccc(C)c(OCCCCc2ccccc2)c1
IUPAC Name 1,4-dimethyl-2-(4-phenylbutoxy)benzene
Molecular Weight (g/mol) 254.367
Formula Weight 254.167
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.