missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethyl-1,2,3,4-tetrahydroquinoxaline-6-carbonitrile, 97%, Thermo Scientific™

Product Code. 10757095
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10757095 1 g 1g
11330607 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10757095 Supplier Thermo Scientific Maybridge Supplier No. CC37116DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 857283-87-5
Molecular Formula C11H13N3
Molecular Weight (g/mol) 187.246
MDL Number MFCD09702356
InChI Key NBANRNWRIHAGBJ-UHFFFAOYSA-N
Synonym 1,4-dimethyl-1,2,3,4-tetrahydroquinoxaline-6-carbonitrile
PubChem CID 24229481
IUPAC Name 1,4-dimethyl-2,3-dihydroquinoxaline-6-carbonitrile
SMILES CN1CCN(C2=C1C=CC(=C2)C#N)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 187.24
Percent Purity 97%
Chemical Name or Material 1,4-Dimethyl-1,2,3,4-tetrahydroquinoxaline-6-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.