missing translation for 'onlineSavingsMsg'
Learn More

(1,4-Dimethyl-1,2,3,4-tetrahydroquinoxalin-6-yl)methylamine, ≥90%, Thermo Scientific™

Product Code. 10065872
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10065872

Brand: Thermo Scientific Maybridge CC37113CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 850375-15-4
Molecular Formula C11H17N3
Molecular Weight (g/mol) 191.278
MDL Number MFCD06797483
InChI Key TWGYATHIWDUKGY-UHFFFAOYSA-N
Synonym 1,4-dimethyl-1,2,3,4-tetrahydroquinoxalin-6-yl methylamine, 1,4-dimethyl-2,3-dihydroquinoxalin-6-yl methanamine, 1,4-dimethyl-1,2,3,4-tetrahydroquinoxalin-6-yl methanamine, 1-1,4-dimethyl-1,2,3,4-tetrahydroquinoxalin-6-yl methanamine, 1,4-dimethyl-1,2,3,4-tetrahydroquinoxalin-6-yl methyl amine, 1-1,4-dimethyl-2,3-dihydroquinoxalin-6-yl methanamine
PubChem CID 7127816
IUPAC Name (1,4-dimethyl-2,3-dihydroquinoxalin-6-yl)methanamine
SMILES CN1CCN(C2=C1C=CC(=C2)CN)C

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 191.28
Percent Purity 90%
Chemical Name or Material (1,4-Dimethyl-1,2,3,4-tetrahydroquinoxalin-6-yl)methylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.