missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethoxybenzene, 99+%

Product Code. 10409220 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10409220 1 kg Glass bottle 1kg
10182710 100 g Glass bottle 100g
10589240 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10409220 Supplier Thermo Scientific Acros Supplier No. 115410010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 150-78-7 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-78-7
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00008401
InChI Key OHBQPCCCRFSCAX-UHFFFAOYSA-N
Synonym p-dimethoxybenzene, p-methoxyanisole, hydroquinone dimethyl ether, benzene, 1,4-dimethoxy, quinol dimethyl ether, benzene, p-dimethoxy, dimethylhydroquinone ether, dimethyl ether hydroquinone, 1,4-dimethoxybenzol, usaf an-9
PubChem CID 9016
IUPAC Name 1,4-dimethoxybenzene
SMILES COC1=CC=C(OC)C=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 54°C to 58°C
Color White
Density 1.0500g/mL
Boiling Point 213°C
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C6H4(OCH3)2
Quantity 1 kg
Beilstein 06, 843
Specific Gravity 1.05
Solubility Information Solubility in water: 0.8g/L (20 c). Other solubilities: soluble in benzene,alcohol,ethylacetate and,acetone,1g/10mL dioxane
Viscosity 1.04 mPa.s (65°C)
Formula Weight 138.17
Percent Purity 99+%
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material 1, 4-Dimethoxybenzene, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.