missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethoxybenzene, 98%

Product Code. 11465203
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465203 2500 g 2500g
11445203 100 g 100g
11455203 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11465203 Supplier Thermo Scientific Alfa Aesar Supplier No. A12822.0E
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 150-78-7 | C8H10O2 | 138.17 g/mol

1,4-Dimethoxybenzene is used as Pharmaceutical Intermediate. It is used in some paints and as a diazo dye. It is also used in perfumes and flavors in order to its floral odor. In addition, it is used on greasy skin, and with sulfur to treat acne, or as a dandruff treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dimethoxybenzene is used as Pharmaceutical Intermediate. It is used in some paints and as a diazo dye. It is also used in perfumes and flavors in order to its floral odor. In addition, it is used on greasy skin, and with sulfur to treat acne, or as a dandruff treatment.

Solubility
Soluble in water(0.83g/L at 20°C) and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-78-7
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00008401
InChI Key OHBQPCCCRFSCAX-UHFFFAOYSA-N
Synonym p-dimethoxybenzene, p-methoxyanisole, hydroquinone dimethyl ether, benzene, 1,4-dimethoxy, quinol dimethyl ether, benzene, p-dimethoxy, dimethylhydroquinone ether, dimethyl ether hydroquinone, 1,4-dimethoxybenzol, usaf an-9
PubChem CID 9016
IUPAC Name 1,4-dimethoxybenzene
SMILES COC1=CC=C(OC)C=C1

Specifications

Melting Point 54°C to 58°C
Density 1.053
Boiling Point 212°C
Flash Point 94°C (201°F)
Odor Sweet
Quantity 2500 g
Beilstein 774605
Sensitivity Light sensitive
Solubility Information Soluble in water(0.83g/L at 20°C) and methanol.
Formula Weight 138.17
Percent Purity 98%
Chemical Name or Material 1,4-Dimethoxybenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.