missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Diisopropylbenzene, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 1,4-Diisopropylbenzene |
| CAS | 100-18-5 |
| Molecular Formula | C12 H18 |
| Quantity | 5 g |
| Synonym | Benzene, p-diisopropyl- (8CI), 1,4-Bis(1-methylethyl)benzene, 1,4-Bis(isopropyl)benzene, 1,4-Diisopropylbenzene, NSC 84198, p-Di-iso-propylbenzene, p-Diisopropylbenzene |
| InChI Formula | InChI=1S/C12H18/c1-9(2)11-5-7-12(8-6-11)10(3)4/h5-10H,1-4H3 |
| SMILES | CC(C)c1ccc(cc1)C(C)C |
| IUPAC Name | 1,4-di(propan-2-yl)benzene |
| Molecular Weight (g/mol) | 162.27 |
| Formula Weight | 162.1409 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction