missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diisopropylbenzene, 99%

Product Code. 10573511 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10573511

Brand: Thermo Scientific Acros 155210250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-18-5 | C12H18 | 162.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-18-5
Molecular Formula C12H18
Molecular Weight (g/mol) 162.28
MDL Number MFCD00008892
InChI Key SPPWGCYEYAMHDT-UHFFFAOYSA-N
Synonym 1,4-diisopropylbenzene, p-diisopropylbenzene, benzene, p-diisopropyl, benzene, 1,4-bis 1-methylethyl, para-diisopropylbenzene, p-diisopropylbenzol, 1,4-bis 1-methylethyl benzene, unii-5m62031nzp, 1,4-di propan-2-yl benzene, dsstox_cid_6652
PubChem CID 7486
SMILES CC(C)C1=CC=C(C=C1)C(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -17.0°C
Density 0.8600g/mL
Boiling Point 203.0°C
Flash Point 76°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H4[CH(CH3)2]2
Refractive Index 1.4890 to 1.4910
Quantity 25 g
Beilstein 05,II,339
Specific Gravity 0.86
Solubility Information Solubility in water: insoluble. Other solubilities: %iscible with alcohol, ether, benzene, acetone and, ccl4
IUPAC Name 1,4-di(propan-2-yl)benzene
Formula Weight 162.27
Percent Purity 99%
Chemical Name or Material 1,4-Diisopropylbenzene, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Diisopropylbenzene
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.