missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diisocyanatobutane, 97%

Product Code. 10318840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10318840 5 g Glass bottle 5g
10173151 1 g Glass bottle 1g
11955681 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10318840 Supplier Thermo Scientific Acros Supplier No. 314380050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4538-37-8 | C6H8N2O2 | 140.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4538-37-8
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.14
MDL Number MFCD00075559
InChI Key OVBFMUAFNIIQAL-UHFFFAOYSA-N
PubChem CID 350453
IUPAC Name 1,4-diisocyanatobutane
SMILES C(CCN=C=O)CN=C=O

Spezifikation

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.1000g/mL
Boiling Point 102.0°C to 104.0°C (14.0 mmHg)
Flash Point 106°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula OCN(CH2)4NCO
Refractive Index 1.4510 to 1.4530
Quantity 5 g
UN Number 2206
Specific Gravity 1.1
Synonym 1,4-butanediisocyanate, tetramethylene diisocyanate, 1,4-disocyanatobutane, acmc-1afua, 1,4-diisocyanato-butane, butane-1,4-diisocyanate, #, butane,1,4-diisocyanato, 1,4-butanediyl diisocyanate, butane, 1,4-diisocyanato
Formula Weight 140.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1, 4-Diisocyanatobutane
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.