missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diiodotetrafluorobenzene, TRC

Product Code. 30377574
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30377574 50 mg
30377232 500 mg
30374352 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30377574 Supplier Toronto Research Chemicals Supplier No. TRCD46607850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 392-57-4
Molecular Formula C6F4I2
Molecular Weight (g/mol) 401.86
IUPAC Name 1,2,4,5-tetrafluoro-3,6-diiodobenzene
SMILES Fc1c(F)c(I)c(F)c(F)c1I

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C6F4I2/c7-1-2(8)6(12)4(10)3(9)5(1)11
Formula Weight 401.8025
Chemical Name or Material 1,4-Diiodotetrafluorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.