missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diiodobenzene, 98%

Product Code. 10137793 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10137793 250 g Glass bottle 250g
10743381 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10137793 Supplier Thermo Scientific Supplier No. 185972500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 624-38-4 | C6H4I2 | 329.9 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-38-4
Molecular Formula C6H4I2
Molecular Weight (g/mol) 329.9
MDL Number MFCD00001054
InChI Key LFMWZTSOMGDDJU-UHFFFAOYSA-N
Synonym p-diiodobenzene, benzene, 1,4-diiodo, p-benzene diiodide, benzene, p-diiodo, unii-i9xcf83vav, i9xcf83vav, 1,4-diiodbenzene, 1,4-diidobenzene, 1,4 diiodobenzene, 1,4-diiodiobenzene
PubChem CID 12208
IUPAC Name 1,4-diiodobenzene
SMILES C1=CC(=CC=C1I)I

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 128°C to 132°C
Color White to Beige
Boiling Point 285°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H4I2
Quantity 250 g
Beilstein 05, 227
Formula Weight 329.9
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 1, 4-Diiodobenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Diiodobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.