missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diiodo-2,5-dimethylbenzene, 97%, Thermo Scientific™

Product Code. 10272532
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10272532 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10272532 Supplier Thermo Scientific Maybridge Supplier No. BTB13663DA
 Restricted Item
Estimated Shipment: 26-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1124-08-9
CAS Min % 96.5
CAS Max % 100.0
Quantity 1 g
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C8H8I2
Linear Formula I2C6H2(CH3)2
MDL Number MFCD00829126
Synonym 2,5-diiodo-p-xylene, benzene,1,4-diiodo-2,5-dimethyl, 2,5-diiodo-rho-xylene, maybridge1_002567, 1,4-bis iodanyl-2,5-dimethyl-benzene
InChI Key WJYYUEQVECTILJ-UHFFFAOYSA-N
SMILES CC1=CC(I)=C(C)C=C1I
IUPAC Name 1,4-diiodo-2,5-dimethylbenzene
Molecular Weight (g/mol) 357.96
PubChem CID 2736171
Formula Weight 357.95
Percent Purity 97%
Chemical Name or Material 1, 4-Diiodo-2, 5-dimethylbenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.