missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dihydroxyanthraquinone, TRC

Product Code. 30135757
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135757 500 mg 500mg
30138196 1 g 1g
30003476 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135757 Supplier Toronto Research Chemicals Supplier No. TRCD455593500MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-64-1
Molecular Formula C14 H8 O4
Molecular Weight (g/mol) 240.21
Synonym Anthraquinone, 1,4-dihydroxy- (8CI), 1,4-Dihydroxy-9,10-anthracenedione, 1,4-Dihydroxy-9,10-anthraquinone, 1,4-Dihydroxyanthraquinone, C.I. 58050, C.I. Solvent Orange 100, C.I. Solvent Orange 86, Chinizarin, DAQ, Macrolex Orange GG, NSC 15367, NSC 229036, NSC 40899, NSC 646569, Quinizarin, Quinizarine, Smoke Orange 18, Smoke Orange R, Solvent Orange 100, Solvent Orange 86, 9,10-Anthracenedione, 1,4-dihydroxy-
IUPAC Name 1,4-dihydroxyanthracene-9,10-dione
SMILES Oc1ccc(O)c2C(=O)c3ccccc3C(=O)c12

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C14H8O4/c15-9-5-6-10(16)12-11(9)13(17)7-3-1-2-4-8(7)14(12)18/h1-6,15-16H
Formula Weight 240.0423
Chemical Name or Material Quinizarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.