missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dihydroxy-2-naphthoic acid, 98%, Thermo Scientific™

Product Code. 15486688
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15486688 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15486688 Supplier Thermo Scientific Alfa Aesar Supplier No. H60210.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1,4-Dihydroxy-2-naphthoic acid from Propionibacterium freudenreichii is known to promote the proliferation of Bifidobacterium. It has potential therapeutic application for psoriasis treatment.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31519-22-9
Molecular Formula C11H8O4
Molecular Weight (g/mol) 204.18
MDL Number MFCD00010370
InChI Key VOJUXHHACRXLTD-UHFFFAOYSA-N
Synonym 1,4-dihydroxy-2-naphthoic acid, 1,4-dihydroxy-2-naphthalenecarboxylic acid, 1,4-dihydroxy-2-naphthoate, 2-naphthalenecarboxylic acid, 1,4-dihydroxy, unii-1u77mr5q7o, 1,4-dihydroxy-2-naphtoic acid, 1,4-dohydroxy-2-naphthoic acid, 1,4-dihydroxy-2-naphthalic acid, cbmicro_022187, acmc-209hn7
PubChem CID 671
ChEBI CHEBI:18094
IUPAC Name 1,4-dihydroxynaphthalene-2-carboxylic acid
SMILES OC(=O)C1=CC(O)=C2C=CC=CC2=C1O

Specifications

Melting Point 220°C (decomposition)
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 204.18
Percent Purity 98%
Chemical Name or Material 1,4-Dihydroxy-2-naphthoic acid
Product Title
Select an issue

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.