missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dihydro-1-methyl-4-oxo-3-pyridinecarboxamide-d3, TRC

Product Code. 30381291
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381291 2.5 mg 2.5mg
30384609 1 mg -
30388752 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381291 Supplier Toronto Research Chemicals Supplier No. TRCD4534172.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1207384-47-1
Molecular Formula C7 2H3 H5 N2 O2
Molecular Weight (g/mol) 155.17
Synonym 1,4-Dihydro-1-(methyl-d3)-4-oxo-3-pyridinecarboxamide
IUPAC Name 4-oxo-1-(trideuteriomethyl)pyridine-3-carboxamide
SMILES [2H]C([2H])([2H])N1C=CC(=O)C(=C1)C(=O)N

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C7H8N2O2/c1-9-3-2-6(10)5(4-9)7(8)11/h2-4H,1H3,(H2,8,11)/i1D3
Formula Weight 155.0774
Percent Purity >95%
Chemical Name or Material N-Methyl-4-pyridone-3-carboxamide-d3
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.