missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diethynylbenzene, 95%

Product Code. p-3757669 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10648904

Brand: Thermo Scientific Acros 391550010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 935-14-8 | C10H6 | 126.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 935-14-8
Molecular Formula C10H6
Molecular Weight (g/mol) 126.16
MDL Number MFCD00078375
InChI Key MVLGANVFCMOJHR-UHFFFAOYSA-N
Synonym p-diethynylbenzene, benzene, 1,4-diethynyl, 1,4-diethynyl-benzene, benzene, diethynyl, benzene,1,4-diethynyl, benzene, p-diethynyl-7ci,8ci, 1,4-bis ethynyl benzene, acmc-209rm8, 4-05-00-01805 beilstein handbook reference
PubChem CID 120463
IUPAC Name 1,4-diethynylbenzene
SMILES C#CC1=CC=C(C=C1)C#C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 94°C to 98°C
Color Brown-Orange or White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C6H4(C∽CH)2
Quantity 1 g
Formula Weight 126.16
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 1, 4-Diethynylbenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Diethynylbenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.