missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diethylpiperazine, 98%

Product Code. 11446446
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446446 250 g 250g
11426446 10 g 10g
11436446 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11446446 Supplier Thermo Scientific Alfa Aesar Supplier No. B24743.30
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6483-50-7 | C8H18N2 | 142.246 g/mol

1,4-Diethylpiperazine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Diethylpiperazine is used as pharmaceutical intermediate.

Solubility
Soluble in methanol, toluene.

Notes
store at a cool and dry place, with container tightly sealed. Store under nitrogen, recommended temperature is below 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6483-50-7
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.246
MDL Number MFCD00126900
InChI Key DDPRYTUJYNYJKV-UHFFFAOYSA-N
PubChem CID 80973
IUPAC Name 1,4-diethylpiperazine
SMILES CCN1CCN(CC1)CC

Specifications

Quantity 250 g
Synonym piperazine, 1,4-diethyl, 1,4-diethyl piperazine, piperazine,1,4-diethyl, acmc-1be2o
Solubility Information Soluble in methanol,toluene.
Formula Weight 142.24
Percent Purity 98%
Chemical Name or Material 1,4-Diethylpiperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.