missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diethylbenzene, 98%

Product Code. 10182520 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10182520 25 mL Glass bottle 25mL
10559240 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10182520 Supplier Thermo Scientific Acros Supplier No. 114150250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 105-05-5 | C10H14 | 134.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-05-5
Molecular Formula C10H14
Molecular Weight (g/mol) 134.22
MDL Number MFCD00009264
InChI Key DSNHSQKRULAAEI-UHFFFAOYSA-N
Synonym p-diethylbenzene, benzene, 1,4-diethyl, p-ethylethylbenzene, benzene, p-diethyl, para-diethylbenzene, unii-0psm16x42d, acmc-1bu3z, dsstox_cid_6711, dsstox_rid_78192, dsstox_gsid_26711
PubChem CID 7734
ChEBI CHEBI:34062
IUPAC Name 1,4-diethylbenzene
SMILES CCC1=CC=C(CC)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -43°C
Color Colorless
Density 0.8600g/mL
Boiling Point 184°C
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H4(C2H5)2
Refractive Index 1.4935 to 1.4955
Quantity 25 mL
Beilstein 05, 426
Specific Gravity 0.86
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in benzene and carbon tetrachloride,miscible with ethanol,ether and acetone
Formula Weight 134.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 4-Diethylbenzene, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Diethylbenzene
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.