missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dicyanobenzene, 98%

Product Code. 10726141 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10726141 100 g Glass bottle 100g
10070840 25 g Glass bottle 25g
10551184 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10726141 Supplier Thermo Scientific Acros Supplier No. 113821000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 623-26-7 | C8H4N2 | 128.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-26-7
Molecular Formula C8H4N2
Molecular Weight (g/mol) 128.13
MDL Number MFCD00001810
InChI Key BHXFKXOIODIUJO-UHFFFAOYSA-N
Synonym terephthalonitrile, 1,4-dicyanobenzene, 1,4-benzenedicarbonitrile, p-phthalodinitrile, 4-cyanobenzonitrile, p-dicyanobenzene, p-benzenedinitrile, p-pdn, terephthalodinitrile, 1,4-benzodinitrile
PubChem CID 12172
IUPAC Name benzene-1,4-dicarbonitrile
SMILES C1=CC(=CC=C1C#N)C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 222°C to 227°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H4(CN)2
Quantity 100 g
Beilstein 09, 846
Solubility Information Solubility in water: 0.08g/L (23°C). Other solubilities: soluble 15.3g/L in acetone (20°C),soluble in methanol
Formula Weight 128.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1, 4-Dicyanobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.