missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichlorophthalazine, 98%, Thermo Scientific Chemicals

Product Code. 15476107
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15476107 5 g 5g
15466107 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476107 Supplier Thermo Scientific Alfa Aesar Supplier No. H33295.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10-7-4752 | C8H4Cl2N2 | 199.03 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10-7-4752
Molecular Formula C8H4Cl2N2
Molecular Weight (g/mol) 199.03
MDL Number MFCD00006909
InChI Key ODCNAEMHGMYADO-UHFFFAOYSA-N
Synonym phthalazine, 1,4-dichloro, ccris 6799, 1,4-dichloro-phthalazine, unii-0ow60266ko, 1,4-dichlorophthalazine, dichlorophthalazine, 1,4-dichloropthalazine, 1,4-dichorophthalazine, pubchem23238, 1,4-dichlorophtalazine
PubChem CID 78490
IUPAC Name 1,4-dichlorophthalazine
SMILES C1=CC=C2C(=C1)C(=NN=C2Cl)Cl

Specifications

Melting Point 164°C to 166°C
Quantity 5 g
Sensitivity Moisture sensitive
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 199.04
Percent Purity 98%
Chemical Name or Material 1,4-Dichlorophthalazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.