missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichlorobenzene, 99+%

Product Code. 11467747
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11467747 2500 g 2500g
11414220 100 g 100g
11457747 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467747 Supplier Thermo Scientific Alfa Aesar Supplier No. A16290.0E
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 106-46-7 | C6H4Cl2 | 146.998 g/mol

1,4-Dichlorobenzene, is used as moth repellant, air fresheners and toilet blocks and general deodorant for garbage and restrom as well as to control fruit borers and ants, may be applied to tobacco seed beds for blue mold control. It is also used as a pharmaceutical intermediate. 1,4-dichlorobenzene is as a used as a minor processing aid in the production of grinding wheels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dichlorobenzene, is used as moth repellant, air fresheners and toilet blocks and general deodorant for garbage and restrom as well as to control fruit borers and ants, may be applied to tobacco seed beds for blue mold control. It is also used as a pharmaceutical intermediate. 1,4-dichlorobenzene is as a used as a minor processing aid in the production of grinding wheels.

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, aluminum and its alloys, some plastics.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-46-7
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 146.998
MDL Number MFCD00000604
InChI Key OCJBOOLMMGQPQU-UHFFFAOYSA-N
Synonym p-dichlorobenzene, paradichlorobenzene, para-dichlorobenzene, paracide, paramoth, dichlorobenzene, paranuggets, santochlor, paradow, evola
PubChem CID 4685
ChEBI CHEBI:28618
IUPAC Name 1,4-dichlorobenzene
SMILES C1=CC(=CC=C1Cl)Cl

Specifications

Melting Point 52°C to 56°C
Density 1.248
Boiling Point 173°C to 174°C
Flash Point 65°C (149°F)
Odor Strong
Quantity 2500 g
UN Number UN3077
Beilstein 1680023
Merck Index 14,3057
Solubility Information It is insoluble in water.
Formula Weight 147
Percent Purity ≥99%
Chemical Name or Material 1,4-Dichlorobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.