missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine, 97%

Product Code. 11363706
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363706 5 g 5g
11353706 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11363706 Supplier Thermo Scientific Alfa Aesar Supplier No. L14835.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 202823-67-4 | C10H10Cl2N2 | 229.104 g/mol

1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202823-67-4
Molecular Formula C10H10Cl2N2
Molecular Weight (g/mol) 229.104
MDL Number MFCD01075667
InChI Key GZIFYLGNTOAIJJ-UHFFFAOYSA-N
Synonym 1,4-dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine, acmc-20ap2h, 5,8-ethanophthalazine, 1,4-dichloro-5,6,7,8-tetrahydro, 3,6-dichloro-4,5-diazatricyclo 6.2.2.0<2,7> dodeca-2 7 ,3,5-triene, 3,6-dichloro-4,5-diazatricyclo 6.2.2.0^ 2,7 dodeca-2,4,6-triene, 1s,8s-3,6-dichloro-4,5-diazatricyclo 6.2.2.0 2 ,? dodeca-2,4,6-triene, 3,6-dichloro-4,5-diazatricyclo 6.2.2.0 2 ,? dodeca-2 7 ,3,5-triene, 3,6-dichloro-4,5-diazatricyclo 6.2.2.0 2 ,? dodeca-2,4,6-triene
PubChem CID 6416107
SMILES C1CC2CCC1C3=C2C(=NN=C3Cl)Cl

Specifications

Melting Point 208°C to 211°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 229.11
Percent Purity 97%
Chemical Name or Material 1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.