missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichloro-2-nitrobenzene, 98%

Product Code. 11413874
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
2500 g
Unit Size:
100g
2500g
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413874 500 g 500g
11403874 100 g 100g
11404080 250 g 250g
11423874 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11413874 Supplier Thermo Scientific Alfa Aesar Supplier No. A11896.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 89-61-2 | C6H3Cl2NO2 | 192.00 g/mol

1,4-Dichloro-2-nitrobenzene is a reagent used in the synthesis of antitrypanosomal, antileishmanial and antimalarial agents. It is also used in the synthesis of lysophosphatidic acid acyltransferase-β in hibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dichloro-2-nitrobenzene is a reagent used in the synthesis of antitrypanosomal, antileishmanial and antimalarial agents. It is also used in the synthesis of lysophosphatidic acid acyltransferase-β in hibitors.

Solubility
Soluble in water, ethanol, ether, benzene, carbon disulfide. Slightly soluble in carbon tetrachloride.

Notes
Incompatible with strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-61-2
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 192.00
MDL Number MFCD00007074
InChI Key RZKKOBGFCAHLCZ-UHFFFAOYSA-N
Synonym 2,5-dichloronitrobenzene, nitro-p-dichlorobenzene, benzene, 1,4-dichloro-2-nitro, 1-nitro-2,5-dichlorobenzene, 2,5-dichlornitrobenzen, 2,5-dichloro-1-nitrobenzene, benzene, 2,5-dichloronitro, 2,5-dichloronitrobenzol, unii-iy18g50ff4, ccris 3096
PubChem CID 6977
IUPAC Name 1,4-dichloro-2-nitrobenzene
SMILES [O-][N+](=O)C1=CC(Cl)=CC=C1Cl

Specifications

Melting Point 53°C to 57°C
Density 1.6
Boiling Point 266°C to 269°C
Flash Point 125°C (257°F)
Quantity 500 g
UN Number UN3077
Beilstein 778109
Solubility Information Soluble in water,ethanol,ether,benzene,carbon disulfide. Slightly soluble in carbon tetrachloride.
Formula Weight 192
Percent Purity 98%
Chemical Name or Material 1,4-Dichloro-2-nitrobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.