Learn More
1,4-Dichloro-2-nitrobenzene, 98%
Description
1,4-Dichloro-2-nitrobenzene is a reagent used in the synthesis of antitrypanosomal, antileishmanial and antimalarial agents. It is also used in the synthesis of lysophosphatidic acid acyltransferase-β in hibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,4-Dichloro-2-nitrobenzene is a reagent used in the synthesis of antitrypanosomal, antileishmanial and antimalarial agents. It is also used in the synthesis of lysophosphatidic acid acyltransferase-β in hibitors.
Solubility
Soluble in water, ethanol, ether, benzene, carbon disulfide. Slightly soluble in carbon tetrachloride.
Notes
Incompatible with strong oxidizing agents, strong bases.
Specifications
Specifications
| Melting Point | 53°C to 57°C |
| Density | 1.6 |
| Boiling Point | 266°C to 269°C |
| Flash Point | 125°C (257°F) |
| Quantity | 250 g |
| UN Number | UN3077 |
| Beilstein | 778109 |
| Solubility Information | Soluble in water,ethanol,ether,benzene,carbon disulfide. Slightly soluble in carbon tetrachloride. |
| Formula Weight | 192 |
| Percent Purity | 98% |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.