missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,4-Dichloro-2-nitrobenzene, 98%

Brand:  Alfa Aesar™ A11896.22

Code : NEW

Additional Details : CAS Number : 89-61-2 Weight : 0.10000kg Transport : UN number : 3077 Chem class : 1 Pack group : III

Quantity 100g

 View more versions of this product

Product Code. 11403874

  • £15.50 / 100g


Estimated Shipment date
from Supplier 23-01-2018
Add to basket

Description

Description

Applications
1,4-Dichloro-2-nitrobenzene is a reagent used in the synthesis of antitrypanosomal, antileishmanial and antimalarial agents. It is also used in the synthesis of lysophosphatidic acid acyltransferase-β in hibitors.

Solubility
Soluble in water, ethanol, ether, benzene, carbon disulfide. Slightly soluble in carbon tetrachloride.

Notes
Incompatible with strong oxidizing agents, strong bases.
Specifications

Chemical Identifiers

1,4-Dichloro-2-nitrobenzene
C6H3Cl2NO2
MFCD00007074
778109
89-61-2
192
UN3077
2,5-Dichloronitrobenzene; Nitro-p-dichlorobenzene

Specifications

1,4-Dichloro-2-nitrobenzene
89-61-2
C6H3Cl2NO2
MFCD00007074
266° to 269°C
53° to 57°C
2,5-Dichloronitrobenzene; Nitro-p-dichlorobenzene
100g
98%
192
UN3077
778109
125°C (257°F)
Videos
SDS