missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromopentane, 97%

Product Code. 11392287
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392287 25 g 25g
11302297 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392287 Supplier Thermo Scientific Alfa Aesar Supplier No. L09842.14
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 626-87-9 | C5H10Br2 | 229.943 g/mol

1,4-Dibromopentane is used in the preparation of pharaoh ant trail pheromone stereoisomer, indolizidine alkaloid. It is also involved in the preparation 1,2-dimethyl-1-phenylcyclopentane and boc-L-2-amino-6-bromoheptanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dibromopentane is used in the preparation of pharaoh ant trail pheromone stereoisomer, indolizidine alkaloid. It is also involved in the preparation 1,2-dimethyl-1-phenylcyclopentane and boc-L-2-amino-6-bromoheptanoic acid.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-87-9
Molecular Formula C5H10Br2
Molecular Weight (g/mol) 229.943
MDL Number MFCD00000153
InChI Key CNBFRBXEGGRSPL-UHFFFAOYSA-N
Synonym pentane, 1,4-dibromo, 1,4-dibrompentan, pentane,4-dibromo, 1,4-dibromo-pentane, pubchem12546, 1,4-dibromopentane, hexanoic acid monoethyl ester
PubChem CID 79082
IUPAC Name 1,4-dibromopentane
SMILES CC(CCCBr)Br

Specifications

Melting Point -34°C
Density 1.687
Boiling Point 102°C to 103°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5085
Quantity 25 g
Beilstein 605295
Solubility Information Insoluble in water.
Formula Weight 229.95
Percent Purity 97%
Chemical Name or Material 1,4-Dibromopentane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.