missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromobutane, 99%

Product Code. p-102509 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
500mL
This item is not returnable. View return policy

Product Code. 10347340

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-52-1 | C4H8Br2 | 215.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-52-1
Molecular Formula C4H8Br2
Molecular Weight (g/mol) 215.92
MDL Number MFCD00000261
InChI Key ULTHEAFYOOPTTB-UHFFFAOYSA-N
Synonym tetramethylene dibromide, butane, 1,4-dibromo, 1,4-butylene bromide, tetramethylene bromide, 1,4-dibrombutan, tetramethylenebromide, alpha,omega-dibromobutane, unii-g49phr6jfu, 1,4-dibrombutan german, 1,4-dibromo-butane
PubChem CID 8056
IUPAC Name 1,4-dibromobutane
SMILES C(CCBr)CBr

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -20°C
Color Brown-Yellow to Colorless
Density 1.8080g/mL
Boiling Point 197°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Br(CH2)4Br
Refractive Index 1.5176 to 1.5196
Quantity 100 mL
Beilstein 01, 120
Fieser 13,313
Specific Gravity 1.808
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in all usual organic solvents and alcohols,soluble in chloroform
Formula Weight 215.92
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1, 4-Dibromobutane, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,4-Dibromobutane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.