missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromobenzene, 98%

Product Code. 11488773
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
2500 g
Unit Size:
100g
2500g
250g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488773 100 g 100g
11434220 50 g 50g
11444220 250 g 250g
11498773 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11488773 Supplier Thermo Scientific Alfa Aesar Supplier No. A10517.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-37-6 | C6H4Br2 | 235.91 g/mol

1,4-Dibromobenzene is used in the synthesis of 2,5-dibromoacetophenone, triarylamines and trans- stilbenes. It is widely used as a heavy liquid solvent as well as a motor oil additive. It finds application as an intermediate in the manufacture of organic chemicals such as dyes, pharmaceuticals and flame retardants for polymeric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dibromobenzene is used in the synthesis of 2,5-dibromoacetophenone, triarylamines and trans- stilbenes. It is widely used as a heavy liquid solvent as well as a motor oil additive. It finds application as an intermediate in the manufacture of organic chemicals such as dyes, pharmaceuticals and flame retardants for polymeric materials.

Solubility
Soluble in alcohol, benzene, chloroform, toluene and carbon tetrachloride. Insoluble in ether.

Notes
Avoid excessive heat and light. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-37-6
Molecular Formula C6H4Br2
Molecular Weight (g/mol) 235.91
MDL Number MFCD00000089
InChI Key SWJPEBQEEAHIGZ-UHFFFAOYSA-N
Synonym p-dibromobenzene, p-bromophenyl bromide, benzene, 1,4-dibromo, benzene, p-dibromo, p-benzene dibromide, 1,4-dibromo benzene, 1,4-dibromo-benzene, unii-9991w3m5hz, dibromobenzene, 1,4, labotest-bb ltbb002071
PubChem CID 7804
ChEBI CHEBI:37150
IUPAC Name 1,4-dibromobenzene
SMILES BrC1=CC=C(Br)C=C1

Specifications

Melting Point 86°C to 90°C
Density 1.841 g/mL
Boiling Point 219°C to 220°C
Flash Point >100°C (212°F)
Assay Percent Range 98%
Refractive Index 1.5742
Quantity 100 g
UN Number UN3077
Beilstein 1904543
Merck Index 14,3019
Solubility Information Soluble in alcohol,benzene,chloroform,toluene and carbon tetrachloride. Insoluble in ether.
Formula Weight 235.91
Percent Purity 98%
Chemical Name or Material 1, 4-Dibromobenzene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,4-Dibromobenzene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1,4-Dibromobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.