missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromo-2-nitrobenzene, 98%

Product Code. 11448703
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448703 25 g 25g
11458703 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11448703 Supplier Thermo Scientific Alfa Aesar Supplier No. A10472.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3460-18-2 | C6H3Br2NO2 | 280.903 g/mol

It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used as an application intermediate. Some of the other applications include HPLC, NMR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used as an application intermediate. Some of the other applications include HPLC, NMR.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from Strong oxidizing agents, Strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3460-18-2
Molecular Formula C6H3Br2NO2
Molecular Weight (g/mol) 280.903
MDL Number MFCD00007046
InChI Key WRGKKASJBOREMB-UHFFFAOYSA-N
Synonym 2,5-dibromonitrobenzene, 1,4-dibromo-2-nitro-benzene, benzene, 1,4-dibromo-2-nitro, 2,5-dibromo-1-nitrobenzene, rarechem fh 1w 0042, pubchem3899, 1,4-dibromonitrobenzene, 2,5-dibromo nitrobenzene, 2,5-dibromo-nitrobenzene, acmc-1ahj1
PubChem CID 77004
IUPAC Name 1,4-dibromo-2-nitrobenzene
SMILES C1=CC(=C(C=C1Br)[N+](=O)[O-])Br

Specifications

Melting Point 82°C to 85°C
Density 2.374
Quantity 25 g
Beilstein 1950425
Solubility Information Slightly soluble in water.
Formula Weight 280.91
Percent Purity 99%
Chemical Name or Material 1,4-Dibromo-2-nitrobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.