missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromo-2-methylbenzene, TRC

Product Code. 30378271
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30378271 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30378271 Supplier Toronto Research Chemicals Supplier No. TRCD425883500MG
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 615-59-8
Molecular Formula C7 H6 Br2
Quantity 500 mg
Synonym Benzene, 1,4-dibromo-2-methyl-, Toluene, 2,5-dibromo- (6CI,7CI,8CI), 1,4-Dibromo-2-methylbenzene, 1,4-Dibromotoluene, 2,5-Dibromotoluene, 3,6-Dibromotoluene, NSC 6222
InChI Formula InChI=1S/C7H6Br2/c1-5-4-6(8)2-3-7(5)9/h2-4H,1H3
SMILES Cc1cc(Br)ccc1Br
IUPAC Name 1,4-dibromo-2-methylbenzene
Molecular Weight (g/mol) 249.93
Formula Weight 247.8836
Chemical Name or Material 2,5-dibromotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.