missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromo-2,5-difluorobenzene, 98%, ACROS Organics™

Product Code. 10175050 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10175050 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10175050 Supplier Thermo Scientific Acros Supplier No. 249500010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 327-51-5
Molecular Formula C6H2Br2F2
Molecular Weight (g/mol) 271.887
MDL Number MFCD00000346
InChI Key GLVMLJCMUBZVTJ-UHFFFAOYSA-N
Synonym 1,4-dibromo-2,5-difluoro-benzene, benzene, 1,4-dibromo-2,5-difluoro, 2,5-difluoro-1,4-dibromo benzene, maybridge1_003946, pubchem3441, acmc-1cmi9, chemwish ic06783, ksc495q1l, emolecules 493137, 2,5-dibromo-1,4-difluorobenzene
PubChem CID 67596
IUPAC Name 1,4-dibromo-2,5-difluorobenzene
SMILES C1=C(C(=CC(=C1Br)F)Br)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 59°C to 63°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula Br2C6H2F2
Quantity 1 g
Formula Weight 271.89
Percent Purity ≥97.5%
Physical Form Adhering Crystals
Chemical Name or Material 1, 4-Dibromo-2, 5-difluorobenzene, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.