missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibenzoylbenzene, 99%, Thermo Scientific™

Product Code. 11499962
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499962 5 g 5g
11409972 25 g 25g
11419972 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499962 Supplier Thermo Scientific Alfa Aesar Supplier No. A12070.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3016-97-5
Molecular Formula C20H14O2
Molecular Weight (g/mol) 286.33
MDL Number MFCD00014087
InChI Key NPENBPVOAXERED-UHFFFAOYSA-N
Synonym 1,4-dibenzoylbenzene, 1,4-phenylenebis phenylmethanone, methanone, 1,4-phenylenebis phenyl, p-dibenzoylbenzene, 4-benzoylbenzophenone, 4-benzoylphenyl phenyl methanone, phenyl 4-phenylcarbonyl phenyl ketone, acmc-1cqof, 4-benzoylphenyl-phenylmethanone
PubChem CID 76395
IUPAC Name (4-benzoylphenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C(=O)C3=CC=CC=C3

Specifications

Melting Point 161°C to 165°C
Quantity 5 g
Beilstein 1880440
Formula Weight 286.33
Percent Purity 99%
Chemical Name or Material 1,4-Dibenzoylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.