missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diazabicyclo[2.2.2]octane, TRC

Product Code. 30355723
Change view
Click to view available options
Quantity:
2.5 g
10 g
100 g
Unit Size:
100g
10g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30355723 2.5 g 2.5g
30367184 100 g 100g
30353755 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30355723 Supplier Toronto Research Chemicals Supplier No. TRCD4171452.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 280-57-9
Molecular Formula C6H12N2
Molecular Weight (g/mol) 112.17
Synonym 1,4-Ethylenepiperazine, 33LSI, 33LV, A 33, A 33LV, AE 33, Activator 105E, Amicure TEDA, Bicyclo[2.2.2]-1,4-diazaoctane, Catalyst A 33, D 33LV, Dabco, Dabco 33, Dabco 33LV, Dabco 33S, Dabco 3LV, Dabco A 33, Dabco Crystal, Dabco Crystalline, Dabco D 0134, Dabco H 1010, Dabco L 1202, Dabco S 25, Jeffcat TD 100, Kaolizer 31, L 33, L 33E, LC 96003, LV 33, Lupragen N 201, Minico L 1020, N,N'-endo-Ethylenepiperazine, NSC 56362, Niax A 33, PC CAT TD 33, PC-TD, PT 301, PT 301 (catalyst), PUR 812, Polycat 33LV, S 25, S 33, TD 100, TED, TEDA, TEDA 33, TEDA 33P, TEDA L 33, TEDA-L 25B, TEDA-L 33B, Tegamine 33, Tego Amine, Tegoamin 33, Texacat TD 100, Texacat TD 33, Thancat TD 33, Thancat TD 33A, Toral SM 2, Toyocat L 33, Toyocat TEDA L 33, Triethylenediamine, 1,4-Ethylenepiperazine, 33LV, A 33, AE 33, Activator 105E, Bicyclo[2.2.2]-1,4-diazaoctane, D 33LV, Dabco, Dabco 33LV, Dabco 3LV, Dabco Crystal, Dabco Crystalline, Dabco L 1202, Dabco S 25, Jeffcat TD 100, Kaolizer 31, L 33, L 33E, LC 96003, LV 33, Minico L 1020, N,N'-endo-Ethylenepiperazine, NSC 56362, Niax A 33, PC CAT TD 33, PC-TD, Polycat 33LV, TD 100, TED, TEDA, Teda L 33, Tegamine 33, Tego Amine, Tegoamin 33, Texacat TD 100, Texacat TD 33, Thancat TD 33, Thancat TD 33A, Toral SM 2, Toyocat L 33, Toyocat TEDA L 33, Triethylenediamine
IUPAC Name 1,4-diazabicyclo[2.2.2]octane
SMILES C1CN2CCN1CC2

Tekniske data

Quantity 2.5 g
InChI Formula InChI=1S/C6H12N2/c1-2-8-5-3-7(1)4-6-8/h1-6H2
Formula Weight 112.1
Chemical Name or Material 1,4-Diazabicyclo[2.2.2]octane
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.