missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diaminobutane dihydrochloride, MedChemExpress

Product Code. 16449426
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16449426 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16449426 Supplier Medchem Express Supplier No. HYY1781

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

MedChemExpress 1,4-Diaminobutane (Putrescine) dihydrochloride is an endogenous metabolite, acts as an indicator of pollution-induced stress in higher plants: barley and rape stressed with Cr(III) or Cr(VI).

1,4-Diaminobutane (Putrescine) dihydrochloride is an endogenous metabolite, acts as an indicator of pollution-induced stress in higher plants: barley and rape stressed with Cr(III) or Cr(VI).

Specifications

Chemical Name or Material 1,4-Diaminobutane dihydrochloride
CAS 333-93-7
Color White
Purity Grade Notes Research
Molecular Formula C4H14Cl2N2
Quantity 100 mg
Synonym Putrescine dihydrochloride
Solubility Information H2O : 50 mg/mL (310.42 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (62.08 mM; Need ultrasonic)
SMILES NCCCCN.[H]Cl.[H]Cl
Molecular Weight (g/mol) 161.07
Formula Weight 161.07
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.