missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Diaminoanthraquinone, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | Disperse Violet 1 |
| CAS | 128-95-0 |
| Molecular Formula | C14H10N2O2 |
| Quantity | 100 g |
| Synonym | 9,10-Anthracenedione, 1,4-diamino-, Anthraquinone, 1,4-diamino- (8CI), Cibacet Violet 2R (6CI), 1,4-Diamino-9,10-anthracenedione, 1,4-Diamino-9,10-anthraquinone, 1,4-Diaminoanthraquinone, Acetate Red Violet R, Acetoquinone Light Heliotrope NL, Acetylon Fast Red Violet R, Akasperse Violet 3R, Amacel Heliotrope R, Amaplast Red Violet P 2R, Artisil Direct Violet 2RP, Artisil Violet 2RP, C.I. 61100, C.I. Disperse Violet 1, C.I. Solvent Violet 11, Celanthrene Red Violet R, Celliton Fast Red Violet, Celliton Fast Red Violet RN, Celliton Fast Red Violet RNA-CF, Celliton Fast Violet R, Celliton Red Violet RN, Celutate Red Violet RH, Cibacet Violet 2RI, Cibacet Violet E 2R, Cilla Fast Red Violet RN, Diacelliton Fast Violet 3R, Diacelliton Fast Violet 5R, Disperse Violet 1, Disperse Violet K, Dispersol Violet B 2RA, Duranol Violet 2R, Fenacet Fast Violet 5R, Gracet Violet 2R, Grasol Violet R, Interchem Acetate Red Violet RRLF, Interchem Acetate Violet R, Interchem Hisperse Violet 2RH, Krisolamine, Microsetile Violet 3R, Miketon Fast Red Violet R, NSC 63807, NSC 7833, Nacelan Violet 4R, Nyloquinone Violet R, Oil Violet R, Oracet Violet 2R, Perliton Violet 3R, Resiren Violet TR, Serilene Brilliant Violet 2R, Serisol Brilliant Violet 2R, Setacyl Violet P-R, Setacyl Violet R, Setile Violet 3R, Smoke Violet LK 6064, Solvent Violet 11, Sudan Violet, Supracet Brilliant Violet 3R, Transetile Violet P 3R, Violet 14447 |
| InChI Formula | InChI=1S/C14H10N2O2/c15-9-5-6-10(16)12-11(9)13(17)7-3-1-2-4-8(7)14(12)18/h1-6H,15-16H2 |
| SMILES | Nc1ccc(N)c2C(=O)c3ccccc3C(=O)c12 |
| IUPAC Name | 1,4-diaminoanthracene-9,10-dione |
| Molecular Weight (g/mol) | 238.24 |
| Formula Weight | 238.0742 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction