missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diaminoanthraquinone, TRC

Product Code. 30353069
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30353069 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30353069 Supplier Toronto Research Chemicals Supplier No. TRCD416080500G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Disperse Violet 1
CAS 128-95-0
Molecular Formula C14H10N2O2
Quantity 500 g
Synonym 9,10-Anthracenedione, 1,4-diamino-, Anthraquinone, 1,4-diamino- (8CI), Cibacet Violet 2R (6CI), 1,4-Diamino-9,10-anthracenedione, 1,4-Diamino-9,10-anthraquinone, 1,4-Diaminoanthraquinone, Acetate Red Violet R, Acetoquinone Light Heliotrope NL, Acetylon Fast Red Violet R, Akasperse Violet 3R, Amacel Heliotrope R, Amaplast Red Violet P 2R, Artisil Direct Violet 2RP, Artisil Violet 2RP, C.I. 61100, C.I. Disperse Violet 1, C.I. Solvent Violet 11, Celanthrene Red Violet R, Celliton Fast Red Violet, Celliton Fast Red Violet RN, Celliton Fast Red Violet RNA-CF, Celliton Fast Violet R, Celliton Red Violet RN, Celutate Red Violet RH, Cibacet Violet 2RI, Cibacet Violet E 2R, Cilla Fast Red Violet RN, Diacelliton Fast Violet 3R, Diacelliton Fast Violet 5R, Disperse Violet 1, Disperse Violet K, Dispersol Violet B 2RA, Duranol Violet 2R, Fenacet Fast Violet 5R, Gracet Violet 2R, Grasol Violet R, Interchem Acetate Red Violet RRLF, Interchem Acetate Violet R, Interchem Hisperse Violet 2RH, Krisolamine, Microsetile Violet 3R, Miketon Fast Red Violet R, NSC 63807, NSC 7833, Nacelan Violet 4R, Nyloquinone Violet R, Oil Violet R, Oracet Violet 2R, Perliton Violet 3R, Resiren Violet TR, Serilene Brilliant Violet 2R, Serisol Brilliant Violet 2R, Setacyl Violet P-R, Setacyl Violet R, Setile Violet 3R, Smoke Violet LK 6064, Solvent Violet 11, Sudan Violet, Supracet Brilliant Violet 3R, Transetile Violet P 3R, Violet 14447
InChI Formula InChI=1S/C14H10N2O2/c15-9-5-6-10(16)12-11(9)13(17)7-3-1-2-4-8(7)14(12)18/h1-6H,15-16H2
SMILES Nc1ccc(N)c2C(=O)c3ccccc3C(=O)c12
IUPAC Name 1,4-diaminoanthracene-9,10-dione
Molecular Weight (g/mol) 238.24
Formula Weight 238.0742
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.