missing translation for 'onlineSavingsMsg'
Learn More

1,4-Di-tert-butylbenzene, 98%

Product Code. 11443367
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443367 5 g 5g
11453367 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11443367 Supplier Thermo Scientific Alfa Aesar Supplier No. A17146.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1012-72-2 | C14H22 | 190.33 g/mol

1,4-di-tert-butylbenzene (DTBB) is used as a crystallized form from several organic solvents to study the effect of solvents and crystallization conditions on its habit. It is also used as a solvent and an intermediate in the manufacture of other organic compounds for the finished products of such as curing agents, engineering plastics and cross-linking agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-di-tert-butylbenzene (DTBB) is used as a crystallized form from several organic solvents to study the effect of solvents and crystallization conditions on its habit. It is also used as a solvent and an intermediate in the manufacture of other organic compounds for the finished products of such as curing agents, engineering plastics and cross-linking agents.

Solubility
Insoluble in water. Soluble in ethanol, benzene, carbon tetrachloride.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1012-72-2
Molecular Formula C14H22
Molecular Weight (g/mol) 190.33
MDL Number MFCD00008836
InChI Key OOWNNCMFKFBNOF-UHFFFAOYSA-N
Synonym 1,4-di-tert-butylbenzene, p-di-tert-butylbenzene, benzene, p-di-tert-butyl, benzene, 1,4-bis 1,1-dimethylethyl, unii-55ptx4kh71, bis 1,1-dimethylethyl benzene, benzene, bis 1,1-dimethylethyl, 1,4-di-t-butylbenzene, acmc-2097up, 1,4-bis tert-butyl benzene
PubChem CID 13895
IUPAC Name 1,4-ditert-butylbenzene
SMILES CC(C)(C)C1=CC=C(C=C1)C(C)(C)C

Specifications

Melting Point 75°C to 79°C
Density 0.985
Boiling Point 236°C
Quantity 5 g
Beilstein 1617000
Solubility Information Insoluble in water. Soluble in ethanol,benzene,carbon tetrachloride.
Formula Weight 190.33
Percent Purity 98%
Chemical Name or Material 1,4-Di-tert-butylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.