missing translation for 'onlineSavingsMsg'
Learn More

1,4-Di-n-butylbenzene, 97+%

Product Code. 15499029
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15499029 10 g 10g
15489029 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499029 Supplier Thermo Scientific Alfa Aesar Supplier No. L16396.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1571-86-4 | C14H22 | 190.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1571-86-4
Molecular Formula C14H22
Molecular Weight (g/mol) 190.33
MDL Number MFCD01076591
InChI Key VGQOZYOOFXEGDA-UHFFFAOYSA-N
Synonym 1,4-di-n-butylbenzene, benzene, 1,4-dibutyl, p-dibutylbenzol, p-dibutylbenzene, acmc-209dfm, p-di-tert-butyl benzene
PubChem CID 519170
IUPAC Name 1,4-dibutylbenzene
SMILES CCCCC1=CC=C(CCCC)C=C1

Specifications

Melting Point -31°C
Density 0.856
Boiling Point 227°C
Flash Point 110°C (230°F)
Refractive Index 1.489
Quantity 10 g
Beilstein 1858055
Formula Weight 190.33
Percent Purity ≥97%
Chemical Name or Material 1,4-Di-n-butylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.