missing translation for 'onlineSavingsMsg'
Learn More

Cerium zirconium oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11393608
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11393608 500 g 500g
11303618 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393608 Supplier Thermo Scientific Alfa Aesar Supplier No. 039216.36
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

Used as wear resistant grinding and dispersion media. Used in fuel cell materials (SOFC), body armors. Also finds applications in catalytic converter, honey comb, precision ceramic parts, abrasive wheels, oxygen sensors, cutting tools. Used as ceramic steel. Other uses include screen printing, dip and spin coating and APS coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53169-24-7
MDL Number MFCD00168089
Synonym Cerium zirconate

Specifications

Physical Form Powder
Quantity 500 g
Solubility Information Soluble in acids. Insoluble in water.
Formula Weight 295.32
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Cerium zirconium oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.