missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanedione monoethylene acetal, 97%

Product Code. 11428603
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428603 5 g 5g
11438603 25 g 25g
11448603 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428603 Supplier Thermo Scientific Alfa Aesar Supplier No. A10408.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4746-97-8 | C8H12O3 | 156.181 g/mol

1,4-Cyclohexanedione is used in the preparation of series of potent analgesic compounds. 1,4-Cyclohexanedione is also used as a building block in the synthesis of tritium labelled probes for the autoradiography study of the dopamine reuptake complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Cyclohexanedione is used in the preparation of series of potent analgesic compounds. 1,4-Cyclohexanedione is also used as a building block in the synthesis of tritium labelled probes for the autoradiography study of the dopamine reuptake complex.

Solubility
Soluble in chloroform & methanol.

Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Incompatible with strong oxidizing agents and strong acids. Ensure adequate ventilation during use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4746-97-8
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.181
MDL Number MFCD00010214
InChI Key VKRKCBWIVLSRBJ-UHFFFAOYSA-N
Synonym 1,4-dioxaspiro 4.5 decan-8-one, 1,4-cyclohexanedione monoethylene acetal, 1,4-cyclohexanedione monoethylene ketal, 1,4-cyclohexanedione monoethyleneacetal, 1,4-dioxa-spiro 4.5 decan-8-one, 1,4-cyclohexanedione monoethyleneketal, cyclohexane-1,4-dione monoethylene ketal, 1,4-dioxaspiro 4.5 decane-8-one
PubChem CID 567415
IUPAC Name 1,4-dioxaspiro[4.5]decan-8-one
SMILES C1CC2(CCC1=O)OCCO2

Specifications

Melting Point 72°C to 75°C
Quantity 5 g
Beilstein 117631
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform & methanol.
Formula Weight 156.18
Percent Purity 97%
Chemical Name or Material 1,4-Cyclohexanedione monoethylene acetal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.