missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Cyclohexanedione Mono(2,2-dimethyltrimethylene Ketal), TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 1,4-Cyclohexanedione Mono(2,2-dimethyltrimethylene Ketal) |
| CAS | 69225-59-8 |
| Molecular Formula | C11H18O3 |
| Quantity | 100 mg |
| Synonym | 3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-one |
| InChI Formula | InChI=1S/C11H18O3/c1-10(2)7-13-11(14-8-10)5-3-9(12)4-6-11/h3-8H2,1-2H3 |
| SMILES | CC1(C)COC2(CCC(=O)CC2)OC1 |
| IUPAC Name | 3,3-dimethyl-1,5-dioxaspiro[5.5]undecan-9-one |
| Molecular Weight (g/mol) | 198.26 |
| Formula Weight | 198.1256 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction